Orders LinkContactUsLinkComponent
Eukitt® mounting medium
Eukitt® mounting medium
Eukitt® mounting medium
Certificaten
2 Opties Hieronder Beschikbaar
Product Details & Documents
This is a mounting medium (contains 45% acrylic resin and 55% xylenes) used in microscopy. It is an adhesive and specimen preservative that can be used manually and in automated coverslipping equipment. Special purpose adhesive and sealant for use with glass, non plastic and other non-reactive materials.
  • Fast-drying (20 minutes)
  • Refractive index close to glass (1.510)
  • Chemically neutral, little sensitivity to water, no reaction to staining
  • Temperature limits: –17 to 42 °C
  • Not affected by ultraviolet light, non yellowing, stays optically clear for over 10 years
  • Spreads quickly and evenly without forming any air bubbles
Can be used in place of epoxy resins for mounting petrographic thin sections, used in fluorescence dyes in bones.
Documents
Specifications for Product Family
Specificaties
Specificaties
  • Catalog No:
  • 03989-100ML
  • 03989-500ML
  • M-Clarity quality level:
  • MQ100
  • MQ100
Specificaties
Product Family Options
Productinformatie
  • Verpakking
    VPE
    Beschikbaarheid
    Price
  • 03989-100ML
    Eukitt® mounting medium
    Eukitt® mounting medium
    Glass bottle
    100 mL
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 100 mL
    1 * 100 mL - 47,10 €
    47,10 €
    1 * 100 mL
       47,10 €
    Specificaties:
    More
    Specifications
    Cat. No.
    03989-100ML
    M-Clarity quality level
    MQ100
  • 03989-500ML
    Eukitt® mounting medium
    Eukitt® mounting medium
    Glass bottle
    500 mL
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 500 mL
    1 * 500 mL - 181,00 €
    181,00 €
    1 * 500 mL
       181,00 €
    Specificaties:
    More
    Specifications
    Cat. No.
    03989-500ML
    M-Clarity quality level
    MQ100
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.