Orders LinkContactUsLinkComponent
Digital timer, three channels
Digital timer, 3 channels, grijs, 65×20×55 mm
  MOIN419CA
 :   MOINEAU
Digital timer, three channels
  MOIN419CA
 :  MOINEAU
Digital timer, three channels
Digital timer, three channels
Digital timer, 3 channels, grijs, 65×20×55 mm
51,70 €
1 * 1 ST
Three-channel (1× count up, 2× count down) timer with metal housing.
  • Timing capacity: 24 hours
  • Two alarms, two memories, snooze function
  • Clock (AM/PM)
 : Supplied with stand, clip, magnet and LR44 battery.
 : Voor installatie, IQOQ, erkende kalibratie onder ISO17025, onderhoud en reparatie mail naar [email protected]
  • Kleur:
    grijs
  • Omschrijving:
    Digital timer, 3 channels
  • b×d×h:
    65×20×55 mm