Stereo zoom microscopes, SZX7
Supplier: OLYMPUS OPTICAL
Certificates
About this item
These modular and scalable stereo zoom microscopes are designed for research applications.
- Apo- and semi-apochromatic objectives
- LED oblique illumination (reflected, transmitted, brightfield, darkfield)
- 'Click-Stop' zoom system for a detachable observations repeatability