Bino and trinocular microscopes, CX41
These microscopes set the standards in both optics and design.
- UIS2 plan achromatic objectives
- Camera can be adapted to trinocular models
- High image clarity: Brightfield, phase contrast, fluorescence
Designed for applications in biology and medicine.
: Supplied with a protective cover and two spare bulbs.
Objectives
Frequently Bought Together