Binocular microscopes, CX22
Supplier: OLYMPUS OPTICAL
Certificates
About this item
This educational microscope with plan achromat objectives and transmitted Köhler illumination ensures an outstanding optical performance thanks to a greater field number (FN of 20) than most standard microscopes in this class.
- LED light source provides long life time of 20000 hours with low power consumption
- Objectives, eyepieces and observation tube with anti-fungus coating
- Moulded grips add comfort and safety when retrieving the microscope from high places, such as shelves
- Eyepieces can be fixed in place to help prevent damage or loss during transportation
- Rotational binocular tube can be locked for secure positioning
- Rackless stage and stage cover for a safe and stable longtime operation
- Focus lock prevents objectives and specimen from any damage
Ideal for use in schools.
Delivery: Supplied ready-to-use with eyepieces, objectives and dust cover.
Frequently Bought Together