Orders LinkContactUsLinkComponent
Electric Sockets and Extension for Bench
Electric Sockets and Extension for Bench
Leverancier:   EQUIP LABO
Electric Sockets and Extension for Bench
Leverancier:  EQUIP LABO
Certificaten
Electric Sockets and Extension for Bench
Electric Sockets and Extension for Bench
2 Opties Hieronder Beschikbaar
Product Details & Documents
For benches, there must be provision for electrical power supplies protected at the top, conforming to the NFC 15 to 100 standard in force, namely one cable for 6×220 V single phase sockets. For the twin pole + earth, 10/16 A it is necessary to provide a 220 V single phase 3G 2,5 to 16 A, 30 mA cable.
Product Family Options
Productinformatie
  • Beschikbaarheid
    Price
  • 132-0044
    Electrical mounting box, 3 sockets
    Electrical mounting box, 3 sockets
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 76,70 €
    76,70 €
    1 * 1 ST
       76,70 €
  • 132-0045
    Electrical mounting box, 2 plugsand 16 A cut out
    Electrical mounting box, 2 plugsand 16 A cut out
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 154,00 €
    154,00 €
    1 * 1 ST
       154,00 €