behrotest® DET supplementary set for fat extraction (100 ml) DFE 100
Catalog # 701-0070
Supplier: BEHR
Specifications
- Type:DFE 100
- Omschrijving:Aanvullingsset voor vetextractie
- VPE:1
Specifications
About this item
This set includes the following equipment:
Reaction vessel 250 ml with flat bottom 65/250
5 Vessel –extractor connector M 2965
5×100 ml fat extractor EZ 100/HAHN
5 Reflux condenser for EZ 100 extractor RFK 100
Ordering information: The DET Basic unit (DBAS, 705-0023) is also required. For other supplementary/expansion sets for DET please contact the Avantor sales office.