Elektrolyse-set, Junior
1 Set
Catalogus # 761-0076
Leverancier: BREUKHOVEN
Ga naar:
Product Details & Documents
- Vermogensafgifte 1 W
- Gasreservoirs: H₂ 20 cm³; O₂ 20 cm³
Specificaties
- Omschrijving:1 Set