Orders LinkContactUsLinkComponent
Terugslagklep
6 - 9 mm ID
Catalogus # 229-3311
Leverancier:   Brand
Terugslagklep
Catalogus # 229-3311
Leverancier:  Brand
Terugslagklep
Terugslagklep
6 - 9 mm ID
Price
4,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
HDPE, transparent. Prevents backflow. With FKM valve disc not suitable for pressure applications.
  • Good chemical resistance
  • Good thermal resistance
  • With tapered ends to ensure that the tube is held firmly
Specificaties
  • Slangafmeting:
    6 - 9 mm ID