Beschermfolie, Bytac
Catalog # 293-4331
Supplier: Saint Gobain Life Sciences
Specifications
- Verpakt:Rol
- Kleur:wit
- l×b:4 600×635 mm
- VPE:4,6 m
Specifications
About this item
Self-adhesive laminate, consisting of a chemical-resistant FEP film surface and a vinyl substrate for protecting all common work surfaces. Can be easily peeled away without leaving any adhesive residue.
Temperature resistance: Vinyl: Up to a maximum of 93 °C ; Aluminium: Up to 182 °C (at a maximum of one hour).