Marker PAP PEN, Immunostaining
720-0169
: MUGEN
:
- VPE:1
- Omschrijving:Markeerstift PAP PEN, immunostainer
PAP PEN is designed to provide a water repellant barrier when a circle is drawn around a specimen. The barrier of the PAP PEN retains antisera within the defined area and ensures, that only the amount of antibody needed for sufficient reaction is used.
- PAP PEN stops spreading, reduces waste and divides slides into discret areas for immunostaining, histology, cytology, cell culture, virology, microbiology
- PAP PEN is effective for immunostaining, PAP method, ABC method, ASD method, enzyme and frozen section methods
- The staining is unsoluble in acetone and alcohol and can be removed if desired by xylene after the staining procedure