Monstername-systeem, Scienceware® Samplit™
Sample jar with scoop, Steriel, 190 ml
Leverancier: Bel-Art Products, a Part of SP
Monstername-systeem, Scienceware® Samplit™
Catalogus # 231-0012
Leverancier: Bel-Art Products, a Part of SP
Ga naar:
Product Details & Documents
PS. This sample collection system comprises a sample jar with integral scoop, allowing samples to be transferred easily and directly to a collection receptacle. Ideally suited for bulk materials, solid, semi-solid, and other materals.
- Available in sterile or non sterile versions
- Designed for rough handling
- Semicircular, extremely robust scoop with peripheral cutting edge
Documents
Certificaten
Zoeken op lotnummerSpecificaties
- Type:Steriel
- Capaciteit:190 ml
- Omschrijving:Sample jar with scoop
- Seq:01