ContactUsLinkComponent
Microtiterplaten, zwart, 96-wells, Sterilin®
Microtiterplaten, zwart, 96-wells, Sterilin®
Catalogus # 634-0006
Leverancier:  STERILIN
undefined
Microtiterplaten, zwart, 96-wells, Sterilin®
Catalogus # 634-0006
Leverancier:  STERILIN
Specificaties
  • Vorm:
    8×12
  • Aant. wells:
    96
  • Steriel:
    -
  • Kleur:
    zwart
  • Omschrijving:
    96 wells plaat, vlakke bodem, zwart
  • VPE:
    50
  • Wellvolume:
    400 µl
Specificaties
Over dit artikel
PS, non-treated. Ideal for fluorescence work and bioluminescence studies.
  • Black 96-well plates are ideal for fluorescence work where the pigment prevents cross-talk between the wells - low background fluorescence minimises light scattering
  • SBS compliant, ensuring suitable fit with all automation
  • Manufactured from virgin polystyrene
  • Supplied nonsterile, providing a medium binding surface
  • Chimney well design to help reduce contamination
  • Supplied with no lid
Kleur: zwart