Orders LinkContactUsLinkComponent
Staining Jar, Coplin
Staining Jar, Coplin
Kleurbakje volgens Coplin
Catalog # 720-0576
Supplier:   SMITH SCIENTIFIC
Staining Jar, Coplin
Catalog # 720-0576
Supplier:  SMITH SCIENTIFIC
Staining Jar, Coplin
Staining Jar, Coplin
Kleurbakje volgens Coplin
Price
40,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Soda-lime glass. With grooves, to accept ten 76×26 mm slides, vertically, back to back.
  • Complete with drop-on lid
Specifications
  • Omschrijving:
    Kleurbakje volgens Coplin