Orders LinkContactUsLinkComponent
Infuusflessen
Infuusflessen
1000 ml
Catalog # 215-9243
Supplier:   MUELLER KREMPEL
Infuusflessen
Catalog # 215-9243
Supplier:  MUELLER KREMPEL
Infuusflessen
Price
8,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Clear glass, without closure.
  • Good chemical and physical properties
  • Screw caps and rubber inserts are available as an option
  • Double relief engraving and standard thread
Specifications
  • Capaciteit:
    1000 ml
Accessories
Product Information
  • Catalog #
    Omschrijving
    Voor
    Availability
    Price
  • MUEL14.076.06
    Rubber discs, grey, 37×4 mm
    1000 ml infusion bottles
    Log in for availability
    1 * 100 ST - 116,00 €
Product Information
  • Catalog #
    Omschrijving
    Voor
    Availability
    Price
  • MUEL14.076.04
    Screw top, stainless steel 18/8, with hole Ø 19 mm
    1000 ml infusion bottles
    Log in for availability
    1 * 100 ST - 143,00 €