Glasvezelfilters
Leverancier: Sartorius
Over dit artikel
These filters are mostly used either for monitoring air and gas or as prefilter. They are manufactured with synthetic binding agents to ensure that the filter has a defined strength.
- Mechanically and chemically stable
- Hydrophilic
23 Opties Hieronder Beschikbaar
Product Details & Documents
These filters are mostly used either for monitoring air and gas or as prefilter. They are manufactured with synthetic binding agents to ensure that the filter has a defined strength.
- Mechanically and chemically stable
- Hydrophilic
Specifications for Product Family
Specificaties
- Catalog No:
- SARTFT-3-01125-045
- SARTFT-3-01125-047
- SARTFT-3-01125-050
- SARTFT-3-01125-110
- SARTFT-3-01125-125
- SARTFT-3-01125-130
- SART13400-13-S
- SART13400-16-S
- SART13400-20-S
- SART13400-25-Q
- 512-0033
- 512-0034
- 512-0035
- 512-0036
- SART13400-80-N
- SART13400-100-K
- SART13430-100-K
- 512-0051
- 512-0053
- 512-0054
- 512-0064
- 512-0055
- 512-0065
- Nr:
- MG 1387/1
- MG 1387/1
- MG 1387/1
- MG 1387/1
- MG 1387/1
- MG 1387/1
- 13400
- 13400
- 13400
- 13400
- 13400
- 13400
- 13400
- 13400
- 13400
- 13400
- 13430
- 13400
- 13400
- 13400
- 13430
- 13400
- 13430
- Environmentally Preferable:
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Lage impact op de fabricage
- Gewicht:
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 223 g/m²
- 74 g/m²
- 74 g/m²
- 74 g/m²
- 223 g/m²
- 74 g/m²
- 223 g/m²
- Dikte:
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 1,30 mm
- 0,34 mm
- 0,34 mm
- 0,34 mm
- 1,30 mm
- 0,34 mm
- 1,30 mm
- Maximumtemperatuur:
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- Milieu certificering:
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
Specificaties
Product Family Options
Productinformatie
- NrMaatBeschikbaarheidPrice
- Specificaties:
More
Specifications
Cat. No.SARTFT-3-01125-045NrMG 1387/1Environmentally PreferableGewicht90 g/m²Dikte0,40 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
More
Specifications
Cat. No.SARTFT-3-01125-047NrMG 1387/1Environmentally PreferableGewicht90 g/m²Dikte0,40 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
More
Specifications
Cat. No.SARTFT-3-01125-050NrMG 1387/1Environmentally PreferableGewicht90 g/m²Dikte0,40 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
More
Specifications
Cat. No.SARTFT-3-01125-110NrMG 1387/1Environmentally PreferableGewicht90 g/m²Dikte0,40 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
More
Specifications
Cat. No.SARTFT-3-01125-125NrMG 1387/1Environmentally PreferableGewicht90 g/m²Dikte0,40 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
More
Specifications
Cat. No.SARTFT-3-01125-130NrMG 1387/1Environmentally PreferableGewicht90 g/m²Dikte0,40 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
More
Specifications
Cat. No.SART13400-13-SNr13400Environmentally PreferableGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.SART13400-16-SNr13400Environmentally PreferableLage impact op de fabricageGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.SART13400-20-SNr13400Environmentally PreferableLage impact op de fabricageGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.SART13400-25-QNr13400Environmentally PreferableLage impact op de fabricageGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.512-0033Nr13400Environmentally PreferableLage impact op de fabricageGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.512-0034Nr13400Environmentally PreferableLage impact op de fabricageGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage - Specificaties:
More
Specifications
Cat. No.512-0035Nr13400Environmentally PreferableGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
More
Specifications
Cat. No.512-0036Nr13400Environmentally PreferableGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.SART13400-80-NNr13400Environmentally PreferableLage impact op de fabricageGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage - Specificaties:
More
Specifications
Cat. No.SART13400-100-KNr13400Environmentally PreferableGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.SART13430-100-KNr13430Environmentally PreferableLage impact op de fabricageGewicht223 g/m²Dikte1,30 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.512-0051Nr13400Environmentally PreferableLage impact op de fabricageGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage - Specificaties:
More
Specifications
Cat. No.512-0053Nr13400Environmentally PreferableGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.512-0054Nr13400Environmentally PreferableLage impact op de fabricageGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.512-0064Nr13430Environmentally PreferableLage impact op de fabricageGewicht223 g/m²Dikte1,30 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage - Specificaties:
More
Specifications
Cat. No.512-0055Nr13400Environmentally PreferableGewicht74 g/m²Dikte0,34 mmMaximumtemperatuur180 °CMilieu certificering - Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.512-0065Nr13430Environmentally PreferableLage impact op de fabricageGewicht223 g/m²Dikte1,30 mmMaximumtemperatuur180 °CMilieu certificeringRenewable Energy CertificateLage impact op de fabricage