Overdrukfilters, AcroCap™, Supor® membraan
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
Supor® hydrophilic polyethersulfone (PES) membrane, acrylic housing: 0,2 µm vent. Positive pressure filtration of aqueous laboratory solutions. For sterilisation or clarification of up to three litres of serum-free cell and tissue culture media, media additives and other aqueous solutions. Ideal for sterilisation of solutions that tend to foam when filtered under vacuum.
- Integral hydrophobic vent prevents air lock
- Reduce mycoplasma using 0,1 µm pore size membrane
- Sterilised by gamma irradiation to eliminate the risk of cytotoxic residuals
Water flow rate at 1 bar: 0,1 µm: 90 ml/min; 0,2 µm: 220 ml/min; 0,45 µm: 500 ml/min.
Inlet/outlet 6,4 mm hose barb, inner accepts male Luer, removable filling bell attached to outlet.
Max. pressure 2,1 bar.
Max. temperature 55 °C.
All products include one filter holder per package.
Frequently Bought Together