Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
Fournisseur:   PRAT DUMAS
Extraction thimbles
Fournisseur:  PRAT DUMAS
Certificates
Extraction thimbles
Extraction thimbles
16 Options Available Below
Product Details & Documents
Pure cotton cellulose
  • Neutral and free of fats and grease
  • Moulded in one piece
  • Very resistant to solvents
Applications include extraction of sludges, asphalt, foodstuffs and in the analysis and quality control of powders and dusts.
Product Family Options
Product Information
  • Ø inw.×h
    Disponibilité
    Prix
  • 80×200 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 711,00 €
    711,00 €
    1 * 25 ST
       711,00 €
  • 80×250 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 782,00 €
    782,00 €
    1 * 25 ST
       782,00 €
  • 58×170 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 434,00 €
    434,00 €
    1 * 25 ST
       434,00 €
  • 46×165 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 198,00 €
    198,00 €
    1 * 25 ST
       198,00 €
  • 41×150 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 179,00 €
    179,00 €
    1 * 25 ST
       179,00 €
  • 46×130 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 198,00 €
    198,00 €
    1 * 25 ST
       198,00 €
  • 34×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 122,00 €
    122,00 €
    1 * 25 ST
       122,00 €
  • 33×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 122,00 €
    122,00 €
    1 * 25 ST
       122,00 €
  • 30×60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 122,00 €
    122,00 €
    1 * 25 ST
       122,00 €
  • 27×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 121,00 €
    121,00 €
    1 * 25 ST
       121,00 €
  • 26×60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 114,00 €
    114,00 €
    1 * 25 ST
       114,00 €
  • 22×60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 114,00 €
    114,00 €
    1 * 25 ST
       114,00 €
  • 70×240 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 538,00 €
    538,00 €
    1 * 25 ST
       538,00 €
  • 37×130 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 136,00 €
    136,00 €
    1 * 25 ST
       136,00 €
  • 30×100 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 122,00 €
    122,00 €
    1 * 25 ST
       122,00 €
  • 22×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 113,00 €
    113,00 €
    1 * 25 ST
       113,00 €