Orders LinkContactUsLinkComponent
Stool and sputum cups
Stool and sputum cups
18 ml
Catalogus # ANICC018S110
Leverancier:   Anicrin
Stool and sputum cups
Catalogus # ANICC018S110
Leverancier:  Anicrin
Stool and sputum cups
Stool and sputum cups
18 ml
Price
214,00 €
Log in voor beschikbaarheid
1 * 1.000 ST
Product Details & Documents
PS, translucent, with white PE stopper
  • Small spoon attached to stopper
  • Good thermal resistance, good chemical resistance
  • Universal application, ideal for single use
Straight-walled containers for sample collection, storage and transport.
Specificaties
  • Capaciteit:
    18 ml