VWR®, Klemmen, driepuntig
Catalog # 241-0015
Supplier: VWR Collection
Specifications
- Spanwijdte:0 - 30 mm
- VPE:5
Specifications
About this item
Aluminiumlegering Laboral.
- Clamping fingers covered in vinyl to protect clamped items
- Easy action knurled screw for adjusting the clamping prongs