Orders LinkContactUsLinkComponent
Verlengkabels
Verlengkabels
PVC extension cable, wit, 3 m
Catalogus # 139-1300
Leverancier:   BACHMANN ELEKTRO
Verlengkabels
Catalogus # 139-1300
Leverancier:  BACHMANN ELEKTRO
Verlengkabels
Verlengkabels
PVC extension cable, wit, 3 m
Price
9,94 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
PVC extension cord with moulded, double-earthed plug and Schuko coupling.
  • Cable (HO5VV-F G31,50 mm²) available in different lengths
  • Colour: white
  • Max. power: 10/16 A, 250 V~
Certifications: VDE compliant
Specificaties
  • Kleur:
    wit
  • Omschrijving:
    PVC extension cable
  • Lengte:
    3 m