Cryobox, System 100™
Catalog # 479-3314
Supplier: Thermo Fisher Scientific
Specifications
- Compartimenten:10×10
- Voor:System 100™ cryobuizen
- Uitw. b×d×h:133×133×52 mm
- Environmentally Preferable:Lage impact op de fabricage
- Milieu certificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- VPE:1
Specifications
About this item
PC, autoclaveerbaar.
Designed for ultra-low temperature storage of System 100™ cryotubes or other similar size, externally or internally threaded vials. For temperatures of –196 to +121 °C.
Transparent lid has printed grid and is keyed to prevent misalignment.