Orders LinkContactUsLinkComponent
MultiScreen® PCR 384 Filter Plates
MultiScreen® PCR 384 Filter Plates
Leverancier:   Merck
MultiScreen® PCR 384 Filter Plates
Leverancier:  Merck
Certificaten
MultiScreen® PCR 384 Filter Plates
2 Opties Hieronder Beschikbaar
Product Details & Documents
  • Colour code: Clear
  • Purification of PCR products
  • Research category
  • Nucleic acid
  • Ambient
Product Family Options
Productinformatie
  • Materiaal
    Wellvolume
    Membraan
    Aant. wells
    Steriel
    Beschikbaarheid
    Price
  • MILFS384PCR10
    MultiScreen® PCR filter plates
    MultiScreen® PCR filter plates
    Polystyreen
    100 μl
    Proprietary membrane
    384
    -
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 10 ST
    1 * 10 ST
  • MILFS384PCR50
    MultiScreen® PCR filter plates
    MultiScreen® PCR filter plates
    Polystyreen
    100 μl
    Proprietary membrane
    384
    -