Captive Spray 2 Emitter
Captivespray 2 emitter pack
Catalogus # BRUK1811112
Leverancier: BRUKER DALTRONIK
Ga naar:
Product Details & Documents
Our ready-to-use fused silica emitter offers a straightforward plug-and-play setup, ensuring hassle-free connection. Available in two internal diameters, our emitters cover both, high-throughput applications and the specific needs of plasma or single-cell proteomics.
- Already attached ZDV union for easier handling
- For use with the captivespray source, captivespray 2 source or captivespray ultra source
The 10 µm diameter is recommended for applications in the low nano flow range (50 to 450 nl/min).
Specificaties
- Omschrijving:Captivespray 2 emitter pack
- Cat. No.:BRUK1811112
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.