RoboSep™-C PBS/EDTA Buffer
RoboSep™-C PBS/EDTA Buffer, 9 L
Catalogus # STMC100-0188
Marketsource
Leverancier: Stemcell Technologies
RoboSep™-C PBS/EDTA Buffer
Catalogus # STMC100-0188
Leverancier: Stemcell Technologies
Marketsource
CAS-nummer:
Ga naar:
Product Details & Documents
PBS buffer containing EDTA.
RoboSep™-C PBS/EDTA Buffer is recommended for use with RoboSep™-C when performing cell separation protocols.
Let op: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
Specificaties
Specificaties
- VPE:9 L
- Omschrijving:RoboSep™-C PBS/EDTA Buffer