Orders LinkContactUsLinkComponent
EasySep™ Mouse CD45 Positive Selection Kit
EasySep™ Mouse CD45 Positive Selection Kit
EasySep™ Mouse CD45 positive selection kit, 2×10⁹ cells
Catalogus # STMC18945
Marketsource
Leverancier:   Stemcell Technologies
EasySep™ Mouse CD45 Positive Selection Kit
Catalogus # STMC18945
Leverancier:  Stemcell Technologies
Marketsource
EasySep™ Mouse CD45 Positive Selection Kit
EasySep™ Mouse CD45 Positive Selection Kit
EasySep™ Mouse CD45 positive selection kit, 2×10⁹ cells
Price
817,00 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
Immunomagnetic positive selection of CD45+ leukocytes from mouse lung, splenocytes, or other tissues.
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Isolated cells are not fluorochrome-labelled
The EasySep™ Mouse CD45 Positive Selection Kit is designed to isolate CD45 cells from single-cell suspensions of splenocytes, lung, or other tissues by positive selection. Desired cells are targeted with antibody complexes recognising CD45 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
Let op: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
Specificaties
  • Omschrijving:
    EasySep™ Mouse CD45 positive selection kit, 2×10⁹ cells