Florisil® for chromatography
Marketsource
Leverancier: Roth Carl
Ga naar:
Over dit artikel
Synoniemen: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
- CAS-nummer:1343-88-0
- Formule:MgO₃Si
- MDL-nummer:MFCD00078249
- Molecular Weight:100.38 g/mol
- EINECS:215-681-1
2 Opties Hieronder Beschikbaar
Product Details & Documents
Synoniemen: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
- CAS-nummer:1343-88-0
- Formule:MgO₃Si
- MDL-nummer:MFCD00078249
- Molecular Weight:100.38 g/mol
- EINECS:215-681-1