Plastic Oily Waste Can
JUSUECN937FLRD
New Product
- Type:Safety cans
- Capaciteit:53 L
- Kleur:rood
- Omschrijving:Plastic oily waste can, hands-free, 937FL, 22×18×21
- Materiaal:HDPE
- VPE:1
Safe collection and storage of oily waste materials, constructed of high density polyethylene (HDPE) and offer fire safe temporary storage of solvent rags, waste rags, shavings and other materials subject to spontaneous combustion.
- Heavy-wall HDPE construction will not rust or dent and is resistant to most chemical compounds for increased durability
- Self-closing lid reduces oxygen preventing a fire from starting
- No seam, leak proof design with molded handles for easy handling
- Feature self-closing lid with foot lever and with a flared base for added stability
- Foot pedal provides hands-free opening
- 'Empty every night' label provides convenient reminder
Ideal for temporary storage of solvent rags, waste rags, shavings and other materials subject to spontaneous combustion - keep one at every workstation.
: FM approved and meets OSHA and NFPA Code 30 requirements.