Orders LinkContactUsLinkComponent
Flask, Iodine
Flask, Iodine
250 ml
Catalogus # WEPA019432
Leverancier:   WEPA PUALUS AND THEWALT
Flask, Iodine
Catalogus # WEPA019432
Flask, Iodine
250 ml
Price
68,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Iodine flask with NS, wide neck made of borosilicate glass, with NS and lid stopper.
  • Standard ground joint: 45/40
Specificaties
  • Capaciteit:
    250 ml