Orders LinkContactUsLinkComponent
Cell Navigator™ Cell Plasma Membrane Staining Kit, Deep Red
Cell Navigator™ Cell Plasma Membrane Staining Kit, Deep Red
Cell Navigator™ Cell plasma membrane staining kit, deep red, 500 Tests
Catalogus # STMC100-0834
Marketsource
Leverancier:   Stemcell Technologies
Cell Navigator™ Cell Plasma Membrane Staining Kit, Deep Red
Catalogus # STMC100-0834
Leverancier:  Stemcell Technologies
Marketsource
CAS-nummer:  
Cell Navigator™ Cell Plasma Membrane Staining Kit, Deep Red
Cell Navigator™ Cell Plasma Membrane Staining Kit, Deep Red
Cell Navigator™ Cell plasma membrane staining kit, deep red, 500 Tests
CAS-nummer:  
Price
752,00 €
Log in voor beschikbaarheid
1 * 500 Tests
Product Details & Documents
For staining cell plasma membranes in fixed cells - 500 tests.
Cell Navigator™ Cell plasma membrane staining kits contain a fluorescently labeled dye for staining the plasma membrane in fixed cells. The fluorescently labeled dye stains the plasma membrane without showing cell-type differences as observed with lectins. The dye can be used as a segmentation tool during high-content screening. Cell Navigator™ Cell plasma membrane staining kit, deep red includes Cellpaint™ Deep Red, live cell staining buffer, and DMSO. Cell Navigator™ Cell plasma membrane staining kit, green includes Cellpaint™ Green, Live cell staining buffer, and DMSO.
Let op: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specificaties
Specificaties
  • VPE:
    500 Tests
  • Omschrijving:
    Cell Navigator™ Cell plasma membrane staining kit, deep red
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.