ContactUsLinkComponent
Annexin V Apoptosis Detection Kit with 7-AAD, PE
Annexin V Apoptosis Detection Kit with 7-AAD, PE
Catalogus # STMC100-0337
Leverancier:  STEMCELL Technologies
CAS-nummer:  
undefined
Annexin V Apoptosis Detection Kit with 7-AAD, PE
Catalogus # STMC100-0337
Leverancier:  STEMCELL Technologies
CAS-nummer:  
Specificaties
  • VPE:
    100
  • Omschrijving:
    Annexin V Apoptosis detection kit with 7-AAD, PE
Specificaties
Over dit artikel
Apoptosis staining and detection kit - 100 tests.
The Annexin V Apoptosis Detection Kit includes Annexin V, Annexin V Binding Buffer, and 7-AAD. The kit is used for the combined detection of early-stage cell apoptosis (using Annexin V) and late-stage cell apoptosis or necrosis (using both Annexin V and 7-AAD).
Annexin V belongs to the annexin family of intracellular proteins that bind to membrane phospholipids in a calcium-dependent manner. This dye has a high affinity for phosphatidylserine (PS) that is present in the inner leaflet of the plasma membrane. During early-stage cell apoptosis, PS is translocated from the inner to the outer leaflet of the cell membrane, exposing it to the external environment, where it is detected by Annexin V. This process of PS translocation occurs prior to the loss of membrane integrity. As cells progress through apoptosis and towards necrosis, the cell membrane is compromised and consequently the 7-AAD viability dye passes into the cell. Thus, cells undergoing early apoptosis stain positive for Annexin V and negative for viability dyes, while apoptotic death or necrosis is characterised by positive staining for both Annexin V and 7-AAD.
Let op: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').