Showa DURACoil 386 Cut Resistant Gloves
Leverancier: Showa
Certificaten
Over dit artikel
Don't compromise on flexibility when seeking highly durable cut-resistant gloves.
- Cut-resistant, abrasion-resistant and oil-resistant
- Form-fitting
- Machine washable
- Extra grip
- Flexible and breathable
- Increased dexterity
- Seamless knit
The DURACoil 386 utilises an advanced HPPE reinforced lining to provide cut and abrasion resistance, without sacrificing dexterity. The microporous coating disperses oil to increase grip. Even for tasks on a smaller scale, such as component assembly, the 386 enables a full range of movement and control.
Certificeringen: Cat II. EN 388:2016+A1:2018, 4X42C.
Verpakking: Gloves are packaged 10 pair per polybag, 12 polybags per case (120 total pair per case).
Specificaties
- Catalog No:
- BEST386-10/XXL
- BEST386-9/XL
- BEST386-7/M
- BEST386-8/L
- BEST386-6/S
- Ambidexter/handspecifiek:
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Voor eenmalig gebruik/herbruikbaar:
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Latex/latexvrij:
- Latex
- Latex
- Latex
- Latex
- Latex
- Gepoederd/poedervrij:
- poedervrij
- poedervrij
- poedervrij
- poedervrij
- poedervrij
- Materiaal:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Specificaties
Frequently Bought Together