InfinityLab Quick Change Inline Filter Assembly
Catalog # HEWL5067-1607
Supplier: Agilent
New Product
Specifications
- Ø:Int.Ø: 2,1 mm
- Poriëngrootte:0,2 µm
- Omschrijving:InfinityLab quick change inline filter assembly, stainless steel
- VPE:1
Specifications
About this item
InfinityLab Quick Change inline filter assembly, for UHPLC Columns. Including five filter discs with integrated Quick Turn Fitting for installation in front of the column.
- Application compatibility: Analytical
- Technique: LC, LC and LC/MS, LC/MS