Vasco® OP eco Surgical and Protective Gloves, Sterile
: B. Braun
New Product
Protection against particulate radioactive contamination.
Protein content ≤ 50 μg/g, lower claims are not considered to be reliable given the expected process variation in manufacture and inter-laboratory testing (EN 455-3).
Contains natural rubber latex which may cause allergic reactions including anaphylactic reactions.
- Catalog No:
- BRAU6081334
- BRAU6081332
- Ambidexter/handspecifiek:
- Ambidexter
- Ambidexter
- Latex/latexvrij:
- Latexvrij
- Latexvrij
- Gepoederd/poedervrij:
- poedervrij
- poedervrij
- Materiaal:
- Natuurlijke rubber latex
- Natuurlijke rubber latex
- Manchetstijl:
- Gerold
- Gerold
- Handpalmdikte:
- —
- 89 ± 5 mm
- Sterility:
- Sterile
- Sterile
Frequently Bought Together