HIS Lite™ Cy3 Tris NTA-Ni Complex
Catalog # AATB12620
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- VPE:100 µG
- Omschrijving:HIS Lite™ Cy3 Tris NTA-Ni complex
Specifications
About this item
Cy3-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.