ContactUsLinkComponent
Amplite® Rapid Colorimetric Biotin Quantitation Kit
Amplite® Rapid Colorimetric Biotin Quantitation Kit
Catalogus # AATB5537
Leverancier:  AAT Bioquest
New Product
CAS-nummer:  
Amplite® Rapid Colorimetric Biotin Quantitation Kit
Catalogus # AATB5537
Leverancier:  AAT Bioquest
New Product
CAS-nummer:  
Specificaties
  • VPE:
    100 Tests
Specificaties
Over dit artikel
Optimised to use with Nanodrop. Amplite™ colorimetric biotin quantitation kit is designed to quantify the number of biotins attached to a protein or other macromolecule with a nanodrop spectrometer.
  • Absorption assessed at 500 nm
  • The kit is best used to determine biotin concentration in the range from 2 to 16 µM
  • 12-month shelf life
It is optimised to use with Nanodrop spectrometer.
Levering: Shipped with blue ice.
Let op: Research use only.