Buccutite™ Rapid APC-Cy7 Tandem Antibody Labelling Kit
Catalog # AATB5415
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- VPE:2
- Includes:2 labellings
Specifications
About this item
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ rapid APC-Cy7 tandem antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PE, APC, PerCP and iFluor® tandem dyes.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery information: Shipped with blue ice.
Caution: Research use only.