Wheat Germ Agglutinin, XFD750 Labelled *XFD750 Same Structure to Alexa Fluor® 750*
Catalog # AATB25515
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- VPE:1 mg
- Omschrijving:Wheat germ agglutinin, XFD750 labelled *XFD750 same structure to Alexa Fluor® 750*
Specifications
About this item
XFD750, manufactured by AAT Bioquest, has an identical chemical structure to Alexa Fluor® 750, a trademark of ThermoFisher.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.