Gelite™ Safe DNA Gel Stain *GelRed Replacement, 10000X in Water*
Catalog # AATB17708
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- VPE:0,5 mL
- Omschrijving:Gelite™ safe DNA gel stain *GelRed replacement, 10000X in water*
Specifications
About this item
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
- Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
Caution: For research use only.