Gelite™ Safe DNA Gel Stain *10000X Water Solution*
Catalog # AATB17699
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- VPE:50 µl
- Omschrijving:Gelite™ safe DNA gel stain *10000X water solution*
Specifications
About this item
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
- Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
Caution: For research use only.