XFD514 NHS ester [Same Structure to Alexa Fluor™ 514 NHS ester]
New Product
:
:916.07 g/mol
Frequently Bought Together
VWR®, Serologische pipetten, standaardreeks
: VWR Collection
Serologische pipetten, Stripette™
: Corning
Cryoboxen, 133×133 mm
: TENAK
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
: VWR Collection
Celcultuurflessen, Falcon®
: Corning
VWR®, Cryoboxes, Half Size, Cardboard
: VWR Collection
VWR®, Strips van PCR-buisjes, zonder doppen
: VWR Collection
Laboratoriumschalen
: KARTELL
VWR®, Cryoboxen, karton, 133 × 133 mm
: VWR Collection
Nalgene®, Biotainer® Flessen
: Thermo Fisher Scientific
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Inzetstukken voor Cryo-Coderdoppen
: VWR Collection
Storage tubes, 2D barcoded, Matrix™
: THERMO MATRIX TECHNOLOGIES
[EN]VIAL 55ML CUSTODY CLEAR 19 1 * 450 ST
: Capitol Europe
Petrischalen
: Anicrin