Canada Balsam Glass Jar
NEUB7298-50-50
New Product
- Capaciteit:570 L
- Omschrijving:Canada Balsma glass jar with glass stirring rod
- b×d×h:866×821×2000 mm
- VPE:1
Canada balsam glass, according to Goldschmidt, is used for microscopy and comes with a glass stirring rod.