Natriumchloride
Colourless HDPE can, 1 L
PRLS90339.290
In accordance with the operating procedure: 320 g Sodium Chloride Ref Prolabo 27810 QSP 1000 ml deionised water The solution may contain a precipitate.
: Saline, NaCl, Sea salt, Halite
- :7647-14-5
- :NaCl
- :MFCD00003477
- :58.44 g/mol
- :1,461 °C
- :2,16 g/cm³
- Verpakking:Colourless HDPE can
- VPE:1 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



