Orders LinkContactUsLinkComponent
Urinalysis Using Simulated Urine
Urinalysis Using Simulated Urine
Urinalysis using simulated urine, 1 KIT
Catalogus # EDVOEVT-1513
Leverancier:   EDVOTEK
Urinalysis Using Simulated Urine
Catalogus # EDVOEVT-1513
Leverancier:  EDVOTEK
CAS-nummer:  
Urinalysis Using Simulated Urine
Urinalysis using simulated urine, 1 KIT
CAS-nummer:  
Price
57,90 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
Urinalysis is one of the oldest medical diagnostic tests, and is still one of the most commonly used tests to this day.
In this activity, students will perform a urinalysis on four different samples of simulated urine. Using a standard urinalysis test strip, students will determine the pH of the samples and test for the presence or absence of glucose, protein, and ketones. Students will then analyze the results of the tests to determine a potential diagnosis for each patient. Kit contains an Instruction Manual and enough material for 2 groups of students.
Delivery information: Kit includes 4 Simulated urine samples, 40 ml each: Patient A, Patient B, Patient C, Control, 8 urine test strips, 8 Plastic cups, 2 Color charts.
Specificaties
Specificaties
  • VPE:
    1 KIT
  • Omschrijving:
    Urinalysis using simulated urine
Specificaties