Orders LinkContactUsLinkComponent
MicroThief Powder Sampler
MicroThief Powder Sampler
Supplier:   Sampling Systems
MicroThief Powder Sampler
Supplier:  Sampling Systems
Certificates
MicroThief Powder Sampler
MicroThief Powder Sampler
2 Options Available Below
Product Details & Documents
The MicroThief powder sampler is designed for the sampling of very small volumes of free-flowing powders.
  • Perfect for sampling very low volumes
  • Ideal for unit dose sampling
  • Suitable for free-flowing powders and granules
  • Fast delivery
  • Available from stock
The MicroThief is ideal for taking very low volume samples of free-flowing powders and granules. The sampler is available in different lengths, allowing point samples to be taken from different depths. The MicroThief is made from chemically resistant 316L stainless steel, which has excellent chemical resistance properties.
Product Family Options
Product Information
  • Omschrijving
    Materiaal
    Lengte
    Volume
    Availability
    Price
  • MicroThief powder sampler
    Roestvrij staal 316L
    350 mm
    0.01 - 0.10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 400,00 €
    1 400,00 €
    1 * 1 ST
       1 400,00 €
  • MicroThief powder sampler
    Roestvrij staal 316L
    500 mm
    0.01 - 0.10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 500,00 €
    1 500,00 €
    1 * 1 ST
       1 500,00 €