Order Entry
Belgium
ContactUsLinkComponent
 
Anti-KCNN3 Rabbit Polyclonal Antibody
  PRSI15-323
New Product
Anti-KCNN3 Rabbit Polyclonal Antibody
  PRSI15-323
New Product

 

  • Type antilichaam:
    Primair
  • antigeen symbool:
    KCNN3
  • Clonality:
    Polyklonaal
  • Conjugatie:
    Onconjugeerd
  • Host:
    Rabbit
  • ImmunoFluorescentie:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulier:
    Liquid
  • Gen-ID:
    3782
  • Antigeen synoniemen:
    SKCA3|hSK3|KCa2.3|KCNN3|SK3
  • Modification:
    None
  • Opslagbuffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Opslagtemperatuur:
    Store at −20 °C. Avoid freeze/thaw cycles
  • Concentratie:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogeen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 237-426 of human KCNN3 (NP_740752.1)
  • Purification:
    Affinity purification
  • Verpakking:
    Vial
  • VPE:
    50 µl

 

 

Type: Primary
Antigen: KCNN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat