Wash Solution Concentrate, 20X
20X wash solution concentrate, Bottle, 1×1 L, 1 L
Catalogus # KPLI50-63-04
Leverancier: SERACARE LIFE SCIENCES
Wash Solution Concentrate, 20X
Catalogus # KPLI50-63-04
Leverancier: SERACARE LIFE SCIENCES
CAS-nummer:
Ga naar:
Product Details & Documents
Convenient 20X concentrate containing imidazole-buffered saline and Tween® 20.
- Suitable for ELISA, blotting, and immunohistochemistry applications
This is used a general immunoassay wash buffer.
Specificaties
Specificaties
- Verpakking:Bottle
- VPE:1 L
- Omschrijving:20X wash solution concentrate
- Info VPE:1×1 L