Orders LinkContactUsLinkComponent
Copper ≥99.5%, powder
Glass bottle for solids, 100 g
  STRE93-2999-100G
Marketsource
Copper ≥99.5%, powder
  STRE93-2999-100G
Marketsource
 :  7440-50-8
Copper ≥99.5%, powder
Glass bottle for solids, 100 g
 :  7440-50-8
47,00 €
1 * 100 g
  •  :7440-50-8
  •  :Cu
  •  :MFCD00010965
  •  :63.55 g/mol
  •  :2567 °C (1013 hPa)
  •  :1083 °C
  •  :8,92
  •  :Kamertemperatuur
  •  :231-159-6
  •  :3089
  •  :4.1,II
  • Verpakking:
    Glass bottle for solids
  • VPE:
    100 g