Euro-line Hinged Lid
Catalogus # MULCBECC64
Leverancier: MULTIROIR
Specificaties
- Kleur:grijs
- Omschrijving:Euro-line hinged lid
- Afmetingen:600×400 mm
- Materiaal:PP
- VPE:2
Specificaties
Over dit artikel
Hinged lids allow to secure the lid to the tray. Goods are thus protected during transport or storage.
- Comes with lid and clasps.
- Compatible with Euro-line bins
- Stackable and recyclable
- Food contact quality
- Solid walls
The lid is very easily attached to the tank by means of a clipping. Opening and closing are carried out with two blue clasps.
Temperature of use: –20°C / +50°C.