Anti-GCSFR Rabbit Monoclonal Antibody [clone: E8E7D]
CSTA10424S
- Type antilichaam:Primair
- antigeen symbool:GCSFR
- Clonality:Monoklonaal
- Clone:E8E7D
- Flowcytometrie:Yes
- Host:Rabbit
- ImmunoNeerslag:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequentie:Q99062 A2-R332
- Opslagtemperatuur:−20 °C
- Shipping temperature:Cold
- VPE:100 µl
Type: Primary
Antigen: GCSFR
Clonality: Monoclonal
Clone: E8E7D
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human